Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpcat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145376.5 | NP_663351.3 | 4, 5 | 602 | ||
GenBank mRNA | AB244717.2 | - | 4, 5 | 602 | ||
AK046079.1 | - | 2, 3 | 397 | |||
AK046507.1 | - | 2, 3 | 397 | |||
AK079946.1 | - | 760 | ||||
AK081865.1 | - | 4, 5 | 760 | |||
AK132379.1 | - | 4, 5 | 736 | |||
AK155286.1 | - | 4, 5 | 766 | |||
AK169190.1 | - | 4, 5 | 735 | |||
AK170723.1 | - | 4, 5 | 755 | |||
AK171582.1 | - | 4, 5 | 755 | |||
BC005662.1 | - | 2, 3 | 132 | |||
BC066809.1 | - | 4, 5 | 551 | 30475544 |