Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D15Ertd621e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145959.3 | NP_666071.2 | 20 | 2179 | ||
GenBank mRNA | AK122297.1 | - | 20 | 2029 | ||
AK137324.1 | - | 20 | 2180 | |||
BC033609.1 | - | 12 | 1261 | 5401763 | ||
BC070426.1 | - | 20 | 2165 | 6405871 | ||
DQ228142.1 | - | 20 | 1958 |