Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D15Ertd621e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145959.3 | NP_666071.2 | 13, 14 | 1424 | ||
GenBank mRNA | AK122297.1 | - | 13, 14 | 1274 | ||
AK137324.1 | - | 13, 14 | 1425 | |||
BC033609.1 | - | 5, 6 | 506 | 5401763 | ||
BC070426.1 | - | 13, 14 | 1410 | 6405871 | ||
DQ228142.1 | - | 13, 14 | 1203 |