Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfdp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184706.1 | NP_001171635.1 | 5 | 634 | ||
NM_001184709.1 | NP_001171638.1 | 3 | 258 | |||
NM_001184710.1 | NP_001171639.1 | 2 | 134 | |||
NM_001184711.1 | NP_001171640.1 | 3 | 238 | |||
NM_178667.4 | NP_848782.2 | 6 | 635 | |||
GenBank mRNA | AK029055.1 | - | 5 | 607 | ||
AK033356.1 | - | 3 | 222 | |||
AK053881.1 | - | 5 | 592 | |||
AK079624.1 | - | 6 | 563 | |||
AK158829.1 | - | 2 | 130 | |||
AK162972.1 | - | 5 | 554 | |||
BC092230.1 | - | 3 | 201 | |||
BC141080.1 | - | 4 | 292 | 9055692 | ||
CA980067.1 | - | 3 | 238 | |||
S79780.1 | - | 4 | 317 |