Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wbscr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145218.3 | NP_660253.2 | 2, 3 | 1347 | ||
GenBank mRNA | AB175684.1 | - | 2, 3 | 425 | ||
AF467979.1 | - | 2, 3 | 634 | |||
AK035817.1 | - | 2, 3 | 1101 | |||
AK048758.1 | - | 2, 3 | 1092 | |||
AK051281.1 | - | 2, 3 | 1747 | |||
AK081848.1 | - | 2, 3 | 741 | |||
AK087726.1 | - | 1902 | ||||
BC052469.1 | - | 2, 3 | 743 | 6810287 | ||
BC069177.1 | - | 2, 3 | 1347 | |||
BC158110.1 | - | 2, 3 | 1374 | 9088184 |