Gene Symbol: Wbscr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145218.3 | NP_660253.2 | 1 | 1143 | ||
GenBank mRNA | AB175684.1 | - | 1 | 221 | ||
AF467979.1 | - | 1 | 430 | |||
AK035817.1 | - | 1 | 897 | |||
AK051281.1 | - | 1 | 1543 | |||
AK051382.1 | - | 1 | 1495 | |||
AK081848.1 | - | 1 | 537 | |||
AK087726.1 | - | 1698 | ||||
BC052469.1 | - | 1 | 539 | 6810287 | ||
BC069177.1 | - | 1 | 1143 | |||
BC158110.1 | - | 1 | 1170 | 9088184 |