Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Phf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081080.1 | NP_001074549.1 | 14 | 3829 | ||
GenBank mRNA | AK122230.1 | - | 10 | 1280 | ||
BC099524.1 | - | 8 | 912 | |||
BC137913.1 | - | 14 | 3835 | 8860932 |