Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081139.1 | NP_001074608.1 | 10 | 2863 | ||
GenBank mRNA | AK028691.1 | - | 10 | 2653 | ||
AK046624.1 | - | 10 | 2673 | |||
BC138276.1 | - | 10 | 2668 | 8861295 | ||
BC145228.1 | - | 10 | 2672 | 9053780 | ||
BC145750.1 | - | 10 | 2668 | 40131053 | ||
BC157973.1 | - | 10 | 2668 | 9007295 |