Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178045.4 | NP_835146.3 | 6, 7 | 689 | ||
GenBank mRNA | AK036528.1 | - | 6, 7 | 653 | ||
AK089460.1 | - | 6, 7 | 647 | |||
AK139284.1 | - | 6, 7 | 636 | |||
AK151561.1 | - | 6, 7 | 654 | |||
AK152639.1 | - | 6, 7 | 654 | |||
AK160584.1 | - | 6, 7 | 663 | |||
AK164357.1 | - | 6, 7 | 652 | |||
AK170955.1 | - | 6, 7 | 638 | |||
AK171198.1 | - | 6, 7 | 667 | |||
BC060709.1 | - | 6, 7 | 644 | 6409692 |