Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021441.2 | NP_067416.2 | 11 | 1158 | ||
GenBank mRNA | AK030522.1 | - | 11 | 1125 | ||
AK052250.1 | - | 11 | 1198 | |||
AK087534.1 | - | 11 | 1148 | |||
AK154846.1 | - | 11 | 1142 | |||
BC051944.1 | - | 11 | 1210 | |||
BC058383.1 | - | 11 | 1100 | 6811470 | ||
Y09974.1 | - | 11 | 1285 |