Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dstyk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172516.4 | NP_766104.2 | 3 | 1257 | ||
GenBank mRNA | AK020880.1 | - | 3 | 1116 | ||
AK083696.1 | - | 3 | 1155 | |||
AK132119.1 | - | 3 | 1132 | |||
AK145449.1 | - | 3 | 1102 | |||
AK172954.1 | - | 3 | 1103 | |||
AK210916.1 | - | 1 | 103 | |||
AY208852.1 | - | 3 | 1104 | |||
AY429676.1 | - | 3 | 1143 | |||
BC130226.1 | - | 3 | 1133 | 40141770 |