Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145396.4 | NP_663371.2 | 10 | 1047 | ||
GenBank mRNA | AK028806.1 | - | 10 | 1015 | ||
AK081572.1 | - | 9 | 1105 | |||
AK081966.1 | - | 10 | 1048 | |||
AK083399.1 | - | 10 | 1030 | |||
AK137554.1 | - | 1016 | ||||
AK149940.1 | - | 10 | 1026 | |||
AK163228.1 | - | 10 | 1017 | |||
AK165125.1 | - | 10 | 1017 | |||
AK167127.1 | - | 10 | 1021 | |||
AK180288.1 | - | 1 | 131 | |||
AK183737.1 | - | 2 | 213 | |||
AK212030.1 | - | 1 | 132 | |||
BC019504.1 | - | 10 | 1000 | 4460615 |
Set Membership: |
![]() |