Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145396.4 | NP_663371.2 | 15, 16 | 1750 | ||
GenBank mRNA | AK028806.1 | - | 15, 16 | 1718 | ||
AK081572.1 | - | 14, 15 | 1808 | |||
AK081966.1 | - | 15, 16 | 1751 | |||
AK083399.1 | - | 15, 16 | 1733 | |||
AK137554.1 | - | 1719 | ||||
AK149940.1 | - | 15, 16 | 1729 | |||
AK163228.1 | - | 15, 16 | 1720 | |||
AK165125.1 | - | 15, 16 | 1720 | |||
AK167127.1 | - | 15, 16 | 1724 | |||
AK218521.1 | - | 1, 2 | 98 |
Set Membership: |
![]() |