Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sidt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289668.1 | NP_001276597.1 | 7 | 1072 | ||
NM_172257.4 | NP_758461.1 | 7 | 1072 | |||
GenBank mRNA | AK152371.1 | - | 7 | 1005 | ||
AK155875.1 | - | 7 | 976 | |||
BC023957.1 | - | 7 | 1076 | 5321190 | ||
BC051101.1 | - | 7 | 800 | 5052849 | ||
CK639655.1 | - | 5 | 403 |