Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sidt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289668.1 | NP_001276597.1 | 26 | 2802 | ||
NM_172257.4 | NP_758461.1 | 25 | 2739 | |||
GenBank mRNA | AK155875.1 | - | 25 | 2643 | ||
AK215815.1 | - | 2 | 169 | |||
BC006873.1 | - | 12 | 1076 | |||
BC023957.1 | - | 25 | 2743 | 5321190 | ||
BC051101.1 | - | 26 | 2530 | 5052849 | ||
CF950439.1 | - | 1 | 85 |