Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Lman2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013374.1 | NP_001013392.1 | 2, 3 | 310 | ||
GenBank mRNA | AK158015.1 | - | 2, 3 | 329 | ||
BC027226.1 | - | 2, 3 | 311 | 2654285 | ||
BC046969.1 | - | 2, 3 | 310 | 6313679 |