Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vezt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031848.1 | - | 9 | 1503 | ||
AK036017.1 | - | 9 | 1440 | |||
AK042540.1 | - | 9 | 1434 | |||
BC056428.1 | - | 9 | 1427 | 5716245 |