Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144817.2 | NP_659066.1 | 3 | 251 | ||
GenBank mRNA | AF428262.1 | - | 3 | 199 | ||
AY212936.1 | - | 2 | 131 | |||
BC021840.1 | - | 3 | 200 | 4502479 | ||
BY730299.1 | - | 3 | 252 |