Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec22c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164562.1 | NP_001158034.1 | 7 | 949 | ||
NM_178677.4 | NP_848792.2 | 8 | 1074 | |||
GenBank mRNA | AK029985.1 | - | 8 | 1005 | ||
AK031125.1 | - | 7 | 946 | |||
AK044249.1 | - | 7 | 949 | |||
AK189904.1 | - | 1 | 75 | |||
BC046289.1 | - | 7 | 904 | 6469221 |