Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomgnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289558.1 | NP_001276487.1 | 4 | 2096 | ||
NM_001289559.1 | NP_001276488.1 | 3 | 1996 | |||
NM_001289560.1 | NP_001276489.1 | 4 | 2092 | |||
NM_153540.4 | NP_705768.4 | 2 | 1939 | |||
GenBank mRNA | AK034473.1 | - | 2 | 1925 | ||
AK042747.1 | - | 4 | 2080 | |||
AK054403.1 | - | 3 | 1984 | |||
AK133802.1 | - | 2 | 1922 | |||
BC025056.1 | - | 2 | 1894 | 4947177 | ||
BC030931.1 | - | 2 | 1901 | 4914234 |