Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ginm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145418.4 | NP_663393.1 | 3 | 279 | ||
GenBank mRNA | AK144481.1 | - | 3 | 240 | ||
BC013529.1 | - | 3 | 279 | 4238526 |