Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146017.3 | NP_666129.1 | 10 | 1284 | ||
GenBank mRNA | AK028946.1 | - | 10 | 1284 | ||
AK160577.1 | - | 10 | 1288 | |||
BC023693.1 | - | 10 | 1271 | 5344479 | ||
BC025550.1 | - | 10 | 1229 | 5320737 | ||
BC027245.1 | - | 10 | 1247 | 3602400 | ||
BC027839.1 | - | 6 | 836 | |||
BC031196.1 | - | 10 | 1258 | 5059564 |