Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271356.1 | NP_001258285.1 | 11, 12 | 1918 | ||
NM_001271357.1 | NP_001258286.1 | 11, 12 | 1661 | |||
NM_146018.2 | NP_666130.1 | 11, 12 | 1958 | |||
GenBank mRNA | AK039106.1 | - | 11, 12 | 1745 | ||
AK080933.1 | - | 11, 12 | 1743 | |||
AK136071.1 | - | 11, 12 | 1649 | |||
AK148216.1 | - | 11, 12 | 1712 | |||
AK154040.1 | - | 11, 12 | 1940 | |||
AK161460.1 | - | 11, 12 | 2001 | |||
AK172245.1 | - | 11, 12 | 1716 | |||
AK177301.1 | - | 2, 3 | 240 | |||
BC025820.1 | - | 11, 12 | 1692 | 5099493 |