Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004143.4 | NP_001004143.2 | 9, 10 | 1396 | ||
GenBank mRNA | AK034492.1 | - | 9, 10 | 1371 | ||
AK084022.1 | - | 5, 6 | 554 | |||
AK155071.1 | - | 9, 10 | 1397 | |||
AK160992.1 | - | 9, 10 | 1305 | |||
AK220368.1 | - | 9, 10 | 1357 | |||
BC026701.1 | - | 2, 3 | 168 | |||
BC052540.1 | - | 5, 6 | 691 | |||
BC058419.1 | - | 2, 3 | 215 | |||
BC080737.1 | - | 9, 10 | 1267 | 30431735 |