Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctage5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165253.1 | NP_001158725.1 | 2 | 191 | ||
NM_001165254.1 | NP_001158726.1 | 2 | 190 | |||
NM_146034.3 | NP_666146.3 | 2 | 190 | |||
GenBank mRNA | AK029466.1 | - | 2 | 190 | ||
AK053010.1 | - | 2 | 124 | |||
AK080312.1 | - | 2 | 166 | |||
AK087188.1 | - | 2 | 127 | |||
AK157358.1 | - | 2 | 192 | |||
AK208297.1 | - | 2 | 63 | |||
BC026864.1 | - | 2 | 142 | 2647214 | ||
BQ571563.1 | - | 2 | 190 |