Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183154.3 | NP_898977.2 | 3 | 1531 | ||
GenBank mRNA | AK090334.1 | - | 4 | 1701 | ||
AK129400.1 | - | 3 | 1506 | |||
AK139825.1 | - | 3 | 1552 | |||
AK170954.1 | - | 3 | 1532 | |||
BC050015.1 | - | 2 | 952 | 30016571 |