Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081017.2 | NP_001074486.2 | 44 | 7649 | ||
GenBank mRNA | AB093296.1 | - | 27 | 4737 | ||
AB257853.1 | - | 44 | 7649 | |||
AK033439.1 | - | 7 | 620 | |||
GQ334471.1 | - | 47 | 7128 |