Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thbs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011580.3 | NP_035710.2 | 8 | 1367 | ||
GenBank mRNA | AK080686.1 | - | 8 | 1402 | ||
AK145202.1 | - | 8 | 1397 | |||
AK163092.1 | - | 8 | 1394 | |||
BC020435.1 | - | 8 | 1375 | |||
BC042422.1 | - | 8 | 1369 | 4018176 | ||
BC050917.1 | - | 8 | 1367 | 6405710 | ||
M87276.1 | - | 8 | 1393 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK089315.1 | - | 3022 | Chr. - - [-] on Build |