Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcho2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172591.3 | NP_766179.2 | 15, 16 | 1539 | ||
GenBank mRNA | AK031041.1 | - | 15, 16 | 1542 | ||
AK142282.1 | - | 15, 16 | 1225 | |||
AK164135.1 | - | 16, 17 | 1397 | |||
BC052456.1 | - | 15, 16 | 1236 | 6830607 |