Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011595.2 | NP_035725.1 | 2 | 583 | ||
GenBank mRNA | AB041611.2 | - | 2 | 440 | ||
AK083905.1 | - | 2 | 441 | |||
AK128943.1 | - | 2 | 442 | |||
AK147847.1 | - | 2 | 441 | |||
BC014713.1 | - | 2 | 405 | 4239015 | ||
L19622.1 | - | 2 | 182 | |||
L27424.1 | - | 2 | 452 | |||
Z30970.1 | - | 2 | 457 |