Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tjp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163574.1 | NP_001157046.1 | 19 | 2970 | ||
NM_009386.2 | NP_033412.2 | 19 | 2970 | |||
GenBank mRNA | BC138028.1 | - | 19 | 2741 | 8861047 | |
BC145100.1 | - | 19 | 2741 | |||
D14340.1 | - | 19 | 2965 |