Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tjp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198985.1 | NP_001185914.1 | 12 | 1880 | ||
NM_011597.4 | NP_035727.2 | 12 | 1870 | |||
GenBank mRNA | AF113005.1 | - | 12 | 1728 | ||
AK028494.1 | - | 12 | 1799 | |||
BC034677.1 | - | 12 | 1788 | 3155078 | ||
BC042695.1 | - | 12 | 1772 | |||
CN527767.1 | - | 4 | 413 |