Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021883.2 | NP_068683.1 | 6 | 729 | ||
GenBank mRNA | AK132878.1 | - | 5 | 562 | ||
BC106849.1 | - | 5 | 617 | 40046092 | ||
BC106850.1 | - | 5 | 617 | 40046093 | ||
S76831.1 | - | 5 | 656 |