Gene Symbol: Pcdh17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013753.2 | NP_001013775.2 | 1 | 4276 | ||
GenBank mRNA | AK139275.1 | - | 1 | 2537 | ||
AK142358.1 | - | 1 | 4272 | |||
AK147265.1 | - | 1 | 4278 | |||
AK158668.1 | - | 1 | 1664 | |||
BC079850.1 | - | 1 | 2273 | |||
BC089004.1 | - | 1 | 2165 | 6841033 |