Search Thermo Fisher Scientific
Gene Symbol: Tnfsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009425.2 | NP_033451.1 | 3, 4 | 383 | ||
GenBank mRNA | AK142960.1 | - | 3, 4 | 385 | ||
AK144931.1 | - | 400 | ||||
AK153581.1 | - | 3, 4 | 433 | |||
AK157633.1 | - | 384 | ||||
BC011469.1 | - | 3, 4 | 349 | |||
BC138807.1 | - | 3, 4 | 357 | 8861829 | ||
BC138808.1 | - | 3, 4 | 357 | 8861830 | ||
BY734631.1 | - | 3, 4 | 400 | |||
U37522.1 | - | 3, 4 | 359 |