Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyro3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290800.1 | NP_001277729.1 | 14 | 2420 | ||
NM_019392.2 | NP_062265.2 | 14 | 1956 | |||
GenBank mRNA | AB000828.1 | - | 14 | 1693 | ||
AK134607.1 | - | 6 | 793 | |||
AK141198.1 | - | 14 | 1727 | |||
BC066058.1 | - | 14 | 1956 | 6834015 | ||
BC082325.1 | - | 14 | 1904 | 30356080 | ||
D17393.1 | - | 14 | 2398 | |||
U05683.1 | - | 14 | 1741 | |||
U18342.1 | - | 14 | 1873 | |||
U18343.1 | - | 14 | 2416 | |||
U18933.1 | - | 14 | 1892 | |||
X78103.1 | - | 14 | 1900 |