Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ucp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009464.3 | NP_033490.1 | 4, 5 | 736 | ||
GenBank mRNA | AB008216.1 | - | 3, 4 | 539 | ||
AB010742.1 | - | 4, 5 | 736 | |||
AF019883.1 | - | 1, 2 | 58 | |||
AF030164.1 | - | 3, 4 | 577 | |||
AF032902.1 | - | 4, 5 | 703 | |||
AF053352.1 | - | 4, 5 | 708 | |||
BC055901.1 | - | 3, 4 | 926 | |||
BC098234.1 | - | 2, 3 | 1432 | |||
BC139430.1 | - | 3, 4 | 604 | 8862452 | ||
BC139431.1 | - | 3, 4 | 604 | 8862453 |