Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nipal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145469.5 | NP_663444.2 | 8, 9 | 889 | ||
GenBank mRNA | AK033975.1 | - | 8, 9 | 890 | ||
AK038556.1 | - | 8, 9 | 1031 | |||
AK079085.1 | - | 8, 9 | 1321 | |||
BC016107.1 | - | 8, 9 | 912 | 4921508 | ||
BC030399.1 | - | 8, 9 | 1406 | 4166825 | ||
BC058207.1 | - | 8, 9 | 1055 | 5361417 |