Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145476.2 | NP_663451.2 | 8 | 1102 | ||
GenBank mRNA | AK039627.1 | - | 8 | 1115 | ||
AK146872.1 | - | 8 | 1103 | |||
AK154226.1 | - | 8 | 1124 | |||
AK154602.1 | - | 8 | 1145 | |||
AK155459.1 | - | 8 | 1116 | |||
AK155770.1 | - | 8 | 1145 | |||
AK178601.1 | - | 4 | 327 | |||
BC023106.1 | - | 5 | 497 | 4016550 | ||
BC066009.1 | - | 8 | 1102 | 5708486 |