Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172113.1 | NP_001165584.1 | 19 | 2057 | ||
NM_001172114.1 | NP_001165585.1 | 18 | 1937 | |||
NM_153792.2 | NP_722487.2 | 19 | 2077 | |||
GenBank mRNA | AK088431.1 | - | 18 | 1846 | ||
AK131617.1 | - | 19 | 2018 | |||
BC005649.1 | - | 7 | 689 | |||
BC008598.1 | - | 19 | 1980 | 3499933 | ||
BC031414.1 | - | 2 | 2227 |