Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Adamts10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172619.3 | NP_766207.2 | 4 | 622 | ||
GenBank mRNA | AK034565.1 | - | 4 | 597 | ||
AK034843.1 | - | 3 | 1710 | |||
AK076334.1 | - | 4 | 596 | |||
BC056427.1 | - | 4 | 592 | |||
BC082773.1 | - | 4 | 614 | |||
BC100558.1 | - | 3 | 376 | |||
BC145353.1 | - | 3 | 529 | |||
BC145354.1 | - | 3 | 529 | |||
BC145355.1 | - | 3 | 529 |