- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Npas4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153553.4 | NP_705781.1 | 3 | 506 | ||
GenBank mRNA | AB049835.1 | - | 3 | 401 | ||
AK032607.1 | - | 3 | 506 | |||
AY730724.1 | - | 3 | 533 | |||
BC059815.1 | - | 3 | 851 | |||
BC079544.1 | - | 3 | 851 | |||
BC129861.1 | - | 3 | 442 | 40110003 |