Gene Symbol: Hectd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163471.1 | NP_001156943.1 | 10 | 1148 | ||
NM_172637.3 | NP_766225.2 | 10 | 1148 | |||
GenBank mRNA | AK029549.1 | - | 10 | 1008 | ||
AK142777.1 | - | 8 | 963 | |||
BC137671.1 | - | 10 | 1119 | 8860690 | ||
BC144815.1 | - | 10 | 1119 | 9053363 | ||
BE864574.1 | - | 3 | 195 | |||
CB526486.1 | - | 7 | 601 |