Gene Symbol: Hectd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163471.1 | NP_001156943.1 | 19 | 2112 | ||
NM_172637.3 | NP_766225.2 | 19 | 2109 | |||
GenBank mRNA | AK029549.1 | - | 19 | 1969 | ||
AK035511.1 | - | 5 | 339 | |||
AK142777.1 | - | 17 | 1936 | |||
BC137671.1 | - | 19 | 2083 | 8860690 | ||
BC144815.1 | - | 19 | 2083 | 9053363 |
Gene Symbol: 1500017E21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005275.1 | - | 655 |