Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam178a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081225.1 | NP_001074694.1 | 17 | 3462 | ||
GenBank mRNA | AK028324.1 | - | 13 | 1414 | ||
AK030089.1 | - | 4 | 338 | |||
AK030951.1 | - | 13 | 1778 | |||
AK031688.1 | - | 17 | 3434 | |||
AK032405.1 | - | 13 | 1417 | |||
AK147588.1 | - | 17 | 3935 | |||
AK161809.1 | - | 13 | 1825 | |||
BC060679.1 | - | 13 | 1379 | |||
BC150999.1 | - | 17 | 3315 | 9087912 | ||
BC157937.1 | - | 17 | 3315 | 9088117 | ||
BC172097.1 | - | 17 | 3315 | 9054383 |