Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam178a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081225.1 | NP_001074694.1 | 7 | 2276 | ||
GenBank mRNA | AK028324.1 | - | 3 | 226 | ||
AK030951.1 | - | 3 | 592 | |||
AK031688.1 | - | 7 | 2248 | |||
AK032405.1 | - | 3 | 231 | |||
AK036700.1 | - | 5 | 1853 | |||
AK147588.1 | - | 7 | 2749 | |||
AK161809.1 | - | 3 | 639 | |||
BC060679.1 | - | 3 | 193 | |||
BC150999.1 | - | 7 | 2129 | 9087912 | ||
BC157937.1 | - | 7 | 2129 | 9088117 | ||
BC172097.1 | - | 7 | 2129 | 9054383 |