Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eno4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178689.4 | NP_848804.2 | 13 | 1715 | ||
GenBank mRNA | AK032395.1 | - | 13 | 1718 | ||
AK076682.1 | - | 9 | 895 | |||
BC023285.1 | - | 10 | 1158 | 3984456 |