Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zbtb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009547.2 | NP_033573.1 | 3 | 1044 | ||
GenBank mRNA | AK037725.1 | - | 3 | 1165 | ||
AK082051.1 | - | 3 | 1185 | |||
AK136762.1 | - | 1140 | ||||
AK145517.1 | - | 3 | 839 | |||
AK146201.1 | - | 3 | 844 | |||
BC052017.1 | - | 3 | 1044 | 5697838 | ||
L15325.1 | - | 2 | 775 |