Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Espnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033292.3 | NP_001028464.1 | 9 | 1500 | ||
GenBank mRNA | AK134448.1 | - | 9 | 1502 |