Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbgt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139197.2 | NP_631936.2 | 7 | 983 | ||
GenBank mRNA | AB084789.1 | - | 6 | 732 | ||
AF292399.1 | - | 6 | 712 | |||
BC106831.2 | - | 6 | 712 | 40044790 |